Learn More
Valutek™ Polyethylene Adhesive Mat
Description
- Each removable layer of Valutek™ adhesive mats has a specially developed adhesive coating designed to provide an effective means of reducing contamination by trapping and preventing the transfer of particles.
- 30 sheets per pad and 4 pads per case (a total of 120 layers)
- Each of the numbered layers of the mat peels off easily to reveal a new clean surface (numbered tabs indicate the number of remaining layers)
Specifications
Specifications
| Dimensions (L x W) | 36 x 45 in. |
| Quantity | 30 Sheets/Mat |
| Color | Blue |
| Material | Polyethylene |