Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dynalon™ Polyethylene Bags, 2.25 mil

Catalog No. 14244014 Shop All Dynalab Corporation Products
Change view
Click to view available options
(W x H) 28 x 36 in.; 2.25 mil
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
14-244-014 (W x H) 28 x 36 in.; 2.25 mil
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 14-244-014 Supplier Dynalon™ Supplier No. 7970050001

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

View Alternate Product

Replacement polyethylene bags for use with floor model disposal boxes.

  • Accessory to 12-009-7A, 12-009-08, and 14-174-20
  • Plastic bags measure 28 in x 36 in. with a thickness of 2.25 mil
  • Floor model disposal box replacement bags are not for use with 12-009-8A Burn Up Bin Floor Model

Specifications

Color Clear
Sterility Non-sterile
Width (English) 28 in.
Thickness 2.25 mil
Height (English) 36 in.
Material Polyethylene
Product Type Bag
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.