Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisherbrand™ Polyethylene Cleanroom Clipboards

Catalog No. 18003068 Shop All Fisherbrand Products
Change view
Click to view available options
Color:
Black
Green
Orange
Purple
Red
Teal
Yellow
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
18-003-068 Purple
18-003-067 Black
18-003-071 Green
18-003-072 Orange
18-003-073 Red
18-003-074 Teal
18-003-070 Yellow
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Catalog No. 18-003-068
Add to Cart
Add to Cart
This item is not returnable. View return policy

Polyethylene cleanroom clipboards with poly clip. Available in 9 colors, for 8.5 x 11 in. documents.

  • Durable and minimal
  • Pre-wiped down with IPA
  • ESD-safe

Specifications

Material Polyethylene
Color Purple
For Use With (Equipment) 8.5 x 11 in. documents
Product Type Cleanroom Clipboard
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.