Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Surgical Design™ Polyethylene Dissecting Board

Catalog No. 22079722 Shop All Surgical Design Products
Change view
Click to view available options
Size:
13 x 11 in.
23 x 16 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
22-079-722 13 x 11 in.
22-079-723 23 x 16 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-079-722 Supplier Surgical Design™ Supplier No. 2850212
Add to Cart
Add to Cart

Multipurpose polyethylene dissecting board.

  • Will not swell, warp or crack
  • Dimensional scales permit quick sizing and serve as part of permanent record
  • Rubber feet stabilize working surface and help prevent board from slipping
  • White background contrast with specimens for excellent photographic details
  • Feature a spill-containment border

Specifications

Product Type Dissection Board
Autoclavable Autoclavable
Color White
Material Polyethylene
For Use With (Application) Dissection
Quantity 1
Size 13 x 11 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.