Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyethylene Glycol Monooleyl Ether (n=approx. 10), TCI America™
SDP

Catalog No. P071425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P071425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P071425G Supplier TCI America Supplier No. P071425G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 9004-98-2
Molecular Formula C58H116O21
Molecular Weight (g/mol) 1149.55
MDL Number MFCD00074369
InChI Key TVSFHLHLHUYZNN-KTKRTIGZSA-N
Synonym emulphor, brij 97, oleth-4, emulphor on-870, oleth-20, oleth-23, volpo 20, brij 98, brij 96, emulphor o
PubChem CID 5364713
IUPAC Name (69Z)-3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxaoctaheptacont-69-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

Specifications

Color Yellow
Physical Form Paste at 20°C
Quantity 25 g
Chemical Name or Material Polyethylene Glycol Monooleyl Ether (n=approx. 10)
EINECSNumber (7)-0097
RTECSNumber RK2800000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.