accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 9004-98-2

IUPAC Name: (69Z)-3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxaoctaheptacont-69-en-1-ol
Synonyms: (69Z)-3,6,9,12,15,18,21,24,27,30,33,36,39,42,45,48,51,54,57,60-icosaoxaoctaheptacont-69-en-1-ol
Molecular Weight (g/mol): —
Molecular Formula: —
InChi Key: TVSFHLHLHUYZNN-KTKRTIGZSA-N
SMILES: CCCCCCCC\C=C/CCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO
Molecular Weight (g/mol) 
  • (7)
  • (1)
Quantity 
  • (1)
  • (1)
  • (4)
  • (1)
  • (1)
Physical Form 
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results