Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Polyethylene Isolation Gown
SDP

Catalog No. 19990127
This item is not returnable. View return policy
Catalog No. 19-990-127 Supplier Keystone Safety Supplier No. ISOPEHD
This item is not returnable. View return policy

Apron-style gown offers protection against the highest level of fluids.

  • Features:
    • 100% Latex-free
    • Flame resistant
    • Conforms to California Proposition 65
    • Produced in an ISO 9001 Certified Factory
  • Recommended For:
    • Lab Applications
    • Medical and Dental Industries
    • Food Industry
    • Assembly Work

Specifications

Product Type Isolation Gown
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Size Universal
Gender Unisex
No. of Pockets None
Material Polyethylene
Color White
Closure Type Ties
Disposable Single-use
Sterility Non-sterile
Cuff Style Thumb Loop
Latex Free Yes
Resistance Fluid, Flame
Certifications/Compliance ISO 9001, California Proposition 65
Autoclavable Not Autoclavable
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.