Learn More
High Five™ Polyethylene Shoe Cover
Catalog No. 19167943 Shop All Ansell Products
Change view
Click to view available options
Color:
Blue
Catalog No. Color
19-167-943 Blue
1 options
This item is not returnable. View return policy
Catalog No. 19-167-943 Supplier High Five™ Supplier No. EB0540005B1K
This item is not returnable. View return policy
Ideal for use in damp work areas
  • Short-term protection from contaminants and liquid splash
  • Strong and durable design
  • Serged seams with elastic top
  • Available in white or blue
  • Latex and silicon free
TRUSTED_SUSTAINABILITY
Specifications
Size X-Large
Material Polyethylene
Color Blue
Closure Type Elastic Ankle
Model AS712
Seam Type Serged
Product Type Shoe Covers