Learn More
Justrite™ Polyethylene Tray/Sump Combination for Shelf #29935 or 4 Gallon Safety Cabinet
Shop All Justrite Safety Group ProductsSpecifications
Specifications
| Color | White |
| For Use With (Equipment) | Fits shelf for 4-gallon cabinet |
| Dimensions (L x W) | 337 x 346 mm (13.25 x 13.265 in.) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.