Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polygalacturonic acid, >86%, MP Biomedicals™

Catalog No. ICN10271105 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10271105 5 g
ICN10271125 25 g
ICN10271180 100 g
ICN10271190 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN10271105 Supplier MP Biomedicals, Inc Supplier No. 0210271105
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25990-10-7
Molecular Formula (C6H8O6)n
Molecular Weight (g/mol) 194.14
MDL Number MFCD00131972
InChI Key AEMOLEFTQBMNLQ-BKBMJHBISA-N
Synonym galacturonan, alpha-d-galactopyranuronic acid, calcium pectate, sodium pectate, alpha-d-galacturonic acid, calcium polygalacturonate, pectate, d-galacturonan, sulfated polygalacturonic acid, galacturonate
PubChem CID 445929
ChEBI CHEBI:33885
IUPAC Name (2S,3R,4S,5R,6S)-3,4,5,6-tetrahydroxyoxane-2-carboxylic acid
SMILES O[C@@H]1[C@@H](O)[C@@H](O-*)[C@H](OC1-*)C(O)=O

Specifications

Color Tan to White
Assay Percent Range >86%
Quantity 5 g
Formula Weight (176)n; n=150 to 400
Percent Purity 88.7%
Physical Form Crystalline Powder
Chemical Name or Material Polygalacturonic acid, from Pectin
Recommended Storage Store at Room Temperature (15 to 30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.