Learn More
Polyscience polycool PG -20
Supplier: Polyscience 060320
PolyScience™ PolyScience° polycool PG -20 -20 to 100°C Temperature Range1 gallon (3.8 L)Features:• Propylene glycol is a safer alternative to ethylene glycol.• Lowers freezing point of water to allow circulation at below freezing temperatures.• Mix with distilled water for broadest temperature range.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.