Learn More
Abcam Polygodial, TRPA1 Agonist, >97% Purity, 25 mg
Description
- Molecular Weight: 234.33 Da
- Purity: >97%
- Biological Activity: Selective TRPA1 agonist (EC50 = 400 nM); inhibits mitochondrial ATPase (IC50 = 15 μM)
- Properties: Antinociceptive in vivo; antifungal
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | TRPA1 |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Buffer | 100% polygodial |
| Content And Storage | Store at -20°C. |
| Formulation | 100% Polygodial |
| Quantity | 25 mg |
| For Use With (Application) | TRPA1 agonism, Antinociception research, Mitochondrial ATPase inhibition, Antifungal research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.