Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abcam Polygodial, TRPA1 Agonist, >97% Purity, 5 mg
Description
- Molecular weight: 234.33 Da
- Purity: >97%
- Potency: Selective TRPA1 agonist (EC50 = 400 nM)
- Biological activity: Antinociceptive in vivo; inhibits mitochondrial ATPase (IC50 = 15 μM); antifungal
- Storage: Store at -20°C
- Shipping: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Content And Storage | Store at -20°C. |
| Target Molecule | TRPA1 |
| Buffer | 100% Polygodial |
| Formulation | 100% Polygodial |
| Quantity | 5 mg |
| For Use With (Application) | TRPA1 activation, mitochondrial ATPase inhibition, antifungal studies, antinociceptive studies |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?