Learn More
Description
Available vacuum or nonvacuum and with or without diluent.
Specifications
Specifications
| Product Type | ESR Vacuum Tube with Diluent |
| For Use With (Equipment) | Polymedco Sediten 10-Minute ESR Reader |
| Quantity | 1000 Box |
| Includes | Diluent |
| Certifications/Compliance | US Federal and state regulations |
| Media Type | 3.8% Sodium Citrate |
| pH | 5.65 |
| Format | Clear Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
