Learn More
Cayman Chemical a-Pyrrolidinopentiophenone me
Supplier: Cayman Chemical 140931 MG
A synthetic compound which interacts with BET proteins with high-affinity (Kd 32 5-42 5 nM) disrupts binding of BET proteins with acetylated histones suppressing the expression of specific inflammatory genes involved in sepsis and endotoxic shock
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.