Learn More
Cayman Chemical ANTIBODY GlycItIn 5mg
SDP
Supplier:  Cayman Chemical 141615 MG
Encompass_Preferred
A natural isoflavone which promotes the proliferation of bone marrow stromal cells and osteoblasts and suppresses bone turnover
Catalog No. 50-295-5473