Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyoxyl 40 Stearate, Type II, NF, Spectrum™ Chemical
SDP

Catalog No. 18605357 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
2.5 kg
12 kg
45 kg
113 kg
Packaging:
Fiber Drum
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-605-357 2.5 kg Poly Pail
18-605-358 12 kg Poly Pail
18-605-359 45 kg Fiber Drum
18-605-360 113 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-605-357 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. P11592.5KG
Add to Cart
Add to Cart

P1159, 9004-99-3

Spectrum™ Chemical Polyoxyl 40 Stearate, Type II, NF is commonly used in pharmaceutical formulations as an emulsifier and a surfactant. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 9004-99-3
Molecular Formula C20H40O3
Molecular Weight (g/mol) 328.54
InChI Key RFVNOJDQRGSOEL-UHFFFAOYSA-N
IUPAC Name 2-hydroxyethyl octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCCO

Specifications

Alkalinity To pass test
CAS Max % 1
Packaging Poly Pail
Quantity 2.5 kg
Saponification Value 25-35
Grade NF
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.