Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gibco™ Polypeptone™ Peptone
GREENER_CHOICE

Catalog No. 297108
Change view
Click to view available options
Quantity:
454 g
10 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
29-710-8 10 kg
B11910 454 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 29-710-8 Supplier Gibco™ Supplier No. 297108
Add to Cart
Add to Cart

Pancreatic digest of casein and peptic digest of animal tissue combined in equal parts

A mixture of peptones made up of equal parts of pancreatic digest of casein and peptic digest of animal tissue.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage Storage conditions: 2°C to 25°C.  Store in cool, dry conditions in well-sealed containers.
Cell Type Bacteria, Fungi, Mammalian
Form Powder
Product Type Dehydrated Culture Media
For Use With (Application) Various bacteria, fungi, and mammalian cells
Product Line Polypeptone
Quantity 10 kg
Source Animal Origin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.