Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ Polyphenol Oxidase (Mushrooms)

Catalog No. 89957628 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
25000 Units
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-957-628 25000 Units
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-957-628 Supplier Abnova™ Supplier No. P5260
Add to Cart
Add to Cart

Used for Enzyme

Specifications

For Use With (Application) Enzyme Activity
Molecular Weight (g/mol) 128kDa
Name Polyphenol Oxidase (Mushrooms)
Preparation Method Native protein purified from Mushrooms (Agaricus bisporus)
Quantity 25000 Units
Storage Requirements Store at -20°C.
Regulatory Status RUO
Common Name POLYPHENOL
Assays Polyphenol Oxidase Assay
Biological Activity ≥500 units per mg dry weight. One unit causes an increase in the absorbance at 280nm of 0.001 per minute at 25°C, pH 6.5, using L-tyrosine as substrate.
Species Fungi
Protein Tag None
Expression System Native protein purified from Mushrooms (Agaricus bisporus)
Form Lyophilized
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.