Learn More
United Scientific™ Polypropylene Beaker Set with Handles
SDPSCI_ED
Catalog No. S43714 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Material:
Polypropylene
Catalog No. Material
S43714 Polypropylene
1 options
Catalog No. S43714 Supplier United Scientific™ Supplier No. PJSET5
Add to Cart
Add to Cart
Set includes five polypropylene beakers can be used as a convenient set of plastic pitchers
  • Short form beakers with raised graduations
  • Molded in polypropylene
  • Clear and autoclavable with good chemical resistance
  • Features a convenient pouring spout and handles for easy lifting
  • Set includes one of each 500 mL, 1000 mL, 2000 mL, 3000 mL, and 5000 mL
Specifications
Description Raised Outer Graduations
Capacity (Metric) 500 mL, 1000 mL, 2000 mL, 3000 mL, and 5000 mL
Material Polypropylene
Type Beaker Set
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Scale Molded Scale
Style Handled