Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Justrite™ Polypropylene, Carboy
GSA_VAGREENER_CHOICE

Catalog No. 19830430 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (Metric):
2.5 L
5 L
10 L
40 L
75 L
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
19-830-430 5 L
19-830-429 2.5 L
19-830-431 10 L
19-830-434 40 L
19-830-435 75 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 19-830-430 Supplier Justrite™ Supplier No. 12929
Add to Cart
Add to Cart

For Use With: For storing and dispensing solvents, buffers, reagents and powders.; Autoclavable; Closure Material: Polypropylene; Color: White

  • Interchangeable caps and adapters provide top connection flexibility
  • Rectangular ergonomic design
  • Constructed from autoclavable USP Class VI, FDA grade materials

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Accuracy ±5%
Autoclavable Autoclavable
Capacity (English) 1.3 gal.
Capacity (Metric) 5 L
Closure Material Polypropylene
Closure Size 83 mm
Color White
Dimensions (L x W x H) 221 x 132 x 414 mm
For Use With (Application) For storing and dispensing solvents, buffers, reagents and powders.
Height (English) 16.3 in.
Length (English) 8.7 in.
Material Polypropylene
Product Type Carboy
Shape Rectangular
Width (English) 5.2 in.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.