Learn More
Description
- Durable polypropylene construction: Built from virgin-grade polypropylene with silicone-lined cap for long-lasting use in liquid storage and transportation.
- Autoclavable and temperature resistant: Withstands steam sterilization and temperatures from 0 to 135°C, ideal for scientific and lab applications.
- FDA and USP compliant: FDA 21 CFR 177.1520 and USP 88 Class VI certified for safe use in food, drug, and medical settings.
- Leakproof and shatter resistant: Tested for crack resistance, handle strength, and leak prevention for worry-free handling of liquids.
- Versatile use cases: Perfect for reagent storage, mixing solutions, and transporting liquids in labs, research facilities, and industrial environments.
- Non-sterile.
Specifications
Specifications
| Autoclavable | Autoclavable |
| Capacity (Metric) | 15 L |
| Color | Translucent |
| Material | Polypropylene |
| Mouth Style | Narrow |
| Product Type | Carboy |
| Sterility | Non-sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
