Learn More
Description
- Durable polypropylene design: Made from virgin-grade polypropylene with a silicone-lined cap and integrated spigot for easy dispensing, ideal for liquid storage and transportation.
- Autoclavable up to 135°C: Withstands steam sterilization and temperature ranges from 0-135°C, making it suitable for scientific and lab applications.
- Certified for safety: FDA 21 CFR 177.1520 and USP 88 Class VI compliant, ensuring safe use in food, drug, and medical applications.
- Leakproof tested: Rigorously tested for crack resistance, handle strength, and leak prevention, ensuring secure and worry-free liquid handling.
- Versatile applications: Perfect for reagent storage, liquid mixing, and dispensing in labs, research facilities, industrial environments, and educational settings.
Specifications
Specifications
| Autoclavable | Not Autoclavable |
| Capacity (Metric) | 20 L |
| Color | Translucent |
| Material | Polypropylene |
| Mouth Style | Narrow |
| Product Type | Carboy |
| Sterility | Non-sterile |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
