Learn More
Eppendorf™ Polypropylene Combitips
GREENER_CHOICE
Catalog No. p-8005622 Shop All Eppendorf North America Products
Change view
Click to view available options
Volume (Metric):
0.1 mL, 0.2 mL, 0.5 mL, 1.0 mL, 2.5 mL, 5.0 mL, 10 mL, 25 mL, 50 mL
Catalog No. Volume (Metric)
05-414-201 0.1 mL, 0.2 mL, 0.5 mL, 1.0 mL, 2.5 mL, 5.0 mL, 10 mL, 25 mL, 50 mL
1 options
This item is not returnable. View return policy
Catalog No. 05-414-201 Supplier Eppendorf™ Supplier No. 0030089936
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Meet all the needs of any modern laboratory with these tips. Eppendorf™ Polypropylene Combitips function according to the positive displacement principle and therefore always dispense the correct volume independently of the density of the liquid and its flow characteristics.
  • Ideal system components for all Eppendorf Multipettes
  • The 9 volume sizes (0.1 to 50 mL) offer a maximum range of dispensing volumes with increments as low as 100 nanoliters depending on the Combitip used
  • Elongated Combitips in volumes 2.5 mL, 5.0 mL and 10 mL make it possible to reach the bottom of the most common laboratory vessels
  • Individual color coding facilitates quick identification
  • Unique funnel geometry ensures comfortable handling while preventing damage to gloves
  • Offered in 3 different purity grades to support any application
TRUSTED_SUSTAINABILITY
Specifications
Description Assortment Pack of Combitips Advanced Standard Positive Displacement Pipet Tips
Volume (Metric) 0.1 mL, 0.2 mL, 0.5 mL, 1.0 mL, 2.5 mL, 5.0 mL, 10 mL, 25 mL, 50 mL
Filtered Non-filtered
Sterility Non-sterile
Clean Claims Eppendorf Quality (Non-sterile)
Packaging 1 CombiTips™ advanced of each size (9 total) and Adapter advanced 25 and 50 mL
Quantity 1 Ea.
Product Type Pipette Tips
Includes One each of the 9 Combitip volume sizes, one 25 mL and one 50 mL Combitip adapter
For Use With (Equipment) Eppendorf Multipette
Material Polypropylene