Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Polypropylene Coveralls
SDP

Catalog No. 19161263
Change view
Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-161-263 Large
19-161-261 Small
19-161-262 Medium
19-161-264 X-Large
19-161-265 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 19-161-263 Supplier Keystone Safety Supplier No. CVLNWELRGWHITE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Cleanroom compatible polypropylene coveralls are breathable and latex-free.

A high-quality, low-cost solution to protect your workers against dry particulates, dirt and grime

  • Non-woven, 100 percent spunbond polypropylene construction
  • Elastic wrists and ankles, zipper front and single collar
  • Latex free
  • Flame resistant
  • Single use/disposable

Specifications

Product Type Cleanroom Coveralls
Size Large
Material Polypropylene
Color White
Disposable Single-use
Ankle Style Elastic
Cuff Style Elastic
Neck Style Collared
Closure Type Front Zipper
Sterility Non-sterile
Collar Type Standard fold
Description Elastic wrist and ankle
Latex Free Yes
For Use With (Application) Contamination in cleanroom environments
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.