Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

AirClean™ Systems Polypropylene Horizontal Laminar Flow Clean Bench
SDP

Catalog No. 361004383 Shop All AirClean Systems Products
Change view
Click to view available options
Height (English):
37 in.
41 in.
Height (Metric):
93.98 cm
104.10 cm
Width (English):
48 in.
60 in.
72 in.
96 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Height (English) Height (Metric) Width (English)
36-100-4383 37 in. 93.98 cm 72 in.
36-100-4382 37 in. 93.98 cm 60 in.
36-100-4381 41 in. 104.10 cm 48 in.
36-100-4384 37 in. 93.98 cm 96 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 36-100-4383 Supplier AirClean™ Systems Supplier No. AC6000HLF
Add to Cart
Add to Cart
Request a Quote

For Class 100 (ISO 5) applications, choose an Airclean Polypropylene Horizontal Laminar Flow Clean Bench.

  • UVTech™ microprocessor controller constantly monitors filter conditions
  • Polypropylene construction
  • Seamless, smooth construction minimizes air turbulence
  • Built-in fluorescent lighting
  • All-white surfaces are easy to clean
  • Available in 110/220V versions
  • Supplied fully assembled

Options

  • Custom sturdy carts or stands
  • Cup sink and vacuum fittings
  • Polypropylene base cabinet
  • Ultraviolet light source
  • IV bar

This product will not protect the operator from biological hazards. Do not use this product with biological hazardous material.

  • IV admixture preparation
  • Drug compounding
  • Plant cell culture
  • Tissue culture (non-biological)
  • Media preparation
  • Pharmaceutical procedures
  • Electronic assembly

Certifications

UL listed, CSA Certified

Compliance

Meets ISO 5 and FS209E Class 100 clean bench standards.

Specifications

Filter Type HEPA
Certifications/Compliance ISO 5 and FS209E Class 100
For Use With (Application) For IV admixture preparation, drug compounding, plant cell culture, tissue culture (non-biological), media preparation, pharmaceutical procedures, electronic assembly
Height (English) 37 in.
Height (English) Interior 27 in.
Height (Metric) 93.98 cm
Height (Metric) Interior 68.58 cm
Voltage 110/220 V
Width (English) 72 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.