Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Safety Cleanroom Compatible Polypropylene Lab Coats
SDP

Catalog No. 19168656
Change view
Click to view available options
Closure Type:
Hook and Loop
Snaps
Cuff Style:
Elastic
Knit
Size:
Small
Medium
Large
X-Large
5X-Large
No. of Pockets:
None
16 product options available for selection
Product selection table with 16 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type Cuff Style Size No. of Pockets
19-168-656 Snaps Knit - -
19-168-627 Snaps Knit Small -
19-813-015 Hook and Loop Elastic Small None
19-168-628 Snaps Knit Medium -
19-813-016 Hook and Loop Elastic Medium None
19-168-653 Snaps Knit - -
19-813-017 Hook and Loop Elastic Large None
19-168-654 Snaps Knit X-Large -
19-813-018 Hook and Loop Elastic X-Large None
19-168-655 Snaps Knit - -
19-813-019 Hook and Loop Elastic - None
19-813-020 Hook and Loop Elastic - None
19-168-657 Snaps Knit - -
19-813-021 Hook and Loop Elastic - None
19-168-658 Snaps Knit 5X-Large -
19-813-022 Hook and Loop Elastic - None
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 16 options available.
16 options
This item is not returnable. View return policy
Catalog No. 19-168-656 Supplier Keystone Safety Supplier No. LC3WKNW3XLWHITE
Add to Cart
Add to Cart
This item is not returnable. View return policy

Polypropylene lab coats are made of a breathable fabric that provides economical protection against dry particulates, dirt, and grime.

  • Non-woven 100% Spunbond polypropylene
  • Strong, high tensile strength
  • Lightweight, breathable
  • Cleanroom compatible
  • Low cost solution to protect workers against dry particulates

Specifications

Product Type Controlled Environments Lab Coat
Gender Unisex
Color White
Closure Type Snaps
Cuff Style Knit
Disposable Single-use
Sterility Non-sterile
Latex Free Yes
For Use With (Application) Manufacturing, Food Service, Industrial, Medical, Pharmaceutical
Construction Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.