Learn More
International Enviroguard™ Polypropylene Lab Coats with Three Pockets, Knit Wrist and Collar
Shop All International Enviroguard Systems ProductsDescription
- Polypropylene lab coats are breathable to help keep users cool throughout their workday
- Low-cost protection against dirt, grime, dust, and dry particulates
- Three pockets allow wearers to take small items with them on the go, like eyeglasses or pens
- Possible applications include food processing, pharmaceuticals, warehousing, general cleaning, environmental cleanup, and more
Specifications
Specifications
| Product Type | General Purpose Lab Coat |
| Size | Large |
| Gender | Unisex |
| Material | Polypropylene |
| Color | Navy |
| Closure Type | Snaps |
| Cuff Style | Knit |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Resistance | Particulates |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.