Learn More
International Enviroguard™ Polypropylene Lab Coats, Elastic Wrist, No Pockets
Catalog No. 19472466 Shop All International Enviroguard Systems Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-472-466 Medium
19-472-467 Large
19-472-468 X-Large
19-472-469 2X-Large
19-472-470 3X-Large
19-472-471 4X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 19-472-466 Supplier International Enviroguard™ Supplier No. 2046EM
Add to Cart
Add to Cart
Polypropylene lab coats provide economical, lightweight protection against dirt, grime, and particulates in non-hazardous environments.
  • Polypropylene lab coats protect from dust, dirt, grime, and dry particulates in non-hazardous environments
  • Often used in food processing, pharmaceuticals, laboratories, cleanrooms, or other controlled environments
  • Breathable and lightweight fabric helps keep wearers cool for long hours
  • Elastic wrists help keep sleeves in place to prevent skin exposure
  • Low-cost protection
Specifications
Product Type Controlled Environments Lab Coat
Size Medium
Gender Unisex
Material Polypropylene
Color White
Closure Type Snaps
Cuff Style Elastic
Disposable Single-use
Sterility Non-sterile
Resistance Particulates
No. of Pockets None
Collar Type Standard
Seam Type Serged