Learn More
International Enviroguard™ Polypropylene Lab Coats, Elastic Wrist, No Pockets
Shop All International Enviroguard Systems ProductsDescription
- Polypropylene lab coats protect from dust, dirt, grime, and dry particulates in non-hazardous environments
- Often used in food processing, pharmaceuticals, laboratories, cleanrooms, or other controlled environments
- Breathable and lightweight fabric helps keep wearers cool for long hours
- Elastic wrists help keep sleeves in place to prevent skin exposure
- Low-cost protection
Specifications
Specifications
| Product Type | Controlled Environments Lab Coat |
| Size | 3X-Large |
| Gender | Unisex |
| Material | Polypropylene |
| Color | White |
| Closure Type | Snaps |
| Cuff Style | Elastic |
| Disposable | Single-use |
| Sterility | Non-sterile |
| Resistance | Particulates |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.