Learn More
Tisch Scientific Polypropylene membrane roll stock, 0.22um, 270mm (width) x 9m (length)
SDP
Supplier:  Tisch Scientific RS30229
Encompass_Preferred
Polypropylene membrane roll stock, 0.22um, 270mm (width) x 9m (length) without substrateThis product has not been established as a suitable replacement for N95 mask material or any other PPE application. In addition, please be aware that we are not able to accept any returned products.
Catalog No. 50-225-1487
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.