Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Polypropylene Microcentrifuge Tubes

Catalog No. 07000304 Shop All Greiner Bio-One Products
Change view
Click to view available options
Includes:
N/A
Support skirt
Closure Color:
Natural
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Closure Color
07-000-304 Support skirt White
07-000-789 N/A Natural
07-000-790 Support skirt Natural
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-304 Supplier Greiner Bio-One Supplier No. 717261
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use in safe and long term storage. Greiner Bio-One Polypropylene Microcentrifuge Tubes offer a leak-free performance and minimal non-specific binding.

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • chemical and temperature resistance
  • clarity coupled to a wide thermal range
  • May be centrifuged to 20,000g
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Free of detectable DNase/RNase and human DNA, non-pyrogenic
Closure Color White
Closure Material Medical Polypropylene
Autoclavable Autoclavable
Capacity (Metric) 1.5 mL
Color Translucent
Material Polypropylene (Medical Grade)
Includes Support skirt
No. per Pack 500
Temperature Range (Metric) -80°C to +121°C
Sterility Sterile
Graduated Yes
Closure Type Screw Cap
Bottom Shape Conical
Max. RCF 20,000 x g (Fixed-angle rotor)
Product Type Reaction Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.