Learn More
Greiner Bio-One Polypropylene Microcentrifuge Tubes
Catalog No. p-7075453 Shop All Greiner Bio-One Products
Change view
Click to view available options
Includes:
N/A
Support skirt
Closure Color:
Natural
White
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes Closure Color
07-000-789 N/A Natural
07-000-790 Support skirt Natural
07-000-304 Support skirt White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-789 Supplier Greiner Bio-One Supplier No. 716201
Only null left
Add to Cart
Add to Cart
For use in safe and long term storage. Greiner Bio-One Polypropylene Microcentrifuge Tubes offer a leak-free performance and minimal non-specific binding.
  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • chemical and temperature resistance
  • clarity coupled to a wide thermal range
  • May be centrifuged to 20,000g
TRUSTED_SUSTAINABILITY
Specifications
Cap Style Screw cap
Closure Color Natural
Color Natural
Material Polypropylene
Includes N/A
Volume (Metric) 1.5mL
Sterility Non-sterile