Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Biotix™ Polypropylene Microcentrifuge Tubes

Catalog No. 12111101 Shop All Biotix Products
Change view
Click to view available options
Color:
Clear
Sterility:
Non-sterile
Capacity (Metric):
0.65 mL
Packaging:
1000 Tubes/Bag
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Sterility Capacity (Metric) Packaging
12-111-101 Clear Non-sterile 0.65 mL 1000 Tubes/Bag
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 12-111-101 Supplier Biotix™ Supplier No. MT0065BC
This item is not returnable. View return policy

Tubes with superior clarity for visualization of samples.

  • DNase, RNase and Endotoxin (pyrogen) free
  • Made from medical-grade virgin polypropylene
TRUSTED_SUSTAINABILITY

Specifications

Description NA
Autoclavable Not Autoclavable
Capacity (Metric) 0.65 mL
Color Clear
Material Polypropylene
Packaging 1000 Tubes/Bag
Sterility Non-sterile
Graduated No
Closure Type Attached Snap Cap
Bottom Shape Conical
Max. RCF 20,000 RCF
Diameter (Metric) 6.38 mm
Shape Cylindrical
Length (Metric) 30 mm
Product Type Microcentrifuge Tube
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.