Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Polypropylene Reagent Bottles with Screw Caps
SCI_ED

Catalog No. S39306 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
4 mL
8 mL
15 mL
30 mL
60 mL
125 mL
250 mL
1 L
Mouth:
Narrow
Wide
Quantity:
1 Ea.
10/Pk.
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric) Mouth Quantity
S39306 1 L Narrow 1 Ea.
S39295 4 mL Narrow 1 Ea.
S88423 4 mL Narrow 10/Pk.
S39296 8 mL Narrow 1 Ea.
S39297 15 mL Narrow 1 Ea.
S39298 30 mL Narrow 1 Ea.
S39299 30 mL Wide 1 Ea.
S39300 60 mL Narrow 1 Ea.
S39301 60 mL Wide 1 Ea.
S39302 125 mL Narrow 1 Ea.
S39303 125 mL Wide 1 Ea.
S39304 250 mL Narrow 1 Ea.
S39305 250 mL Wide 1 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. S39306 Supplier Eisco™ Supplier No. CH0172EN
Add to Cart
Edge
Add to Cart

Includes

bottle, threaded screw cap

Transparent, rigid polypropylene reagent bottles with screw caps can be used to store liquid solutions in the laboratory. Threaded screw cap renders the bottles spill and leak proof. Autoclavable. Narrow and wide neck varieties available.

  • Made of high quality, rigid polypropylene (PP)
  • Includes threaded polypropylene screw cap
  • Designed to store liquid solutions in the laboratory
  • Spill and leak resistant
  • Autoclavable
  • Narrow and wide mouth varieties available

Warranty and Services

1 year limited manufacturer warranty

Specifications

Capacity (Metric) 1 L
Color Clear
Description Leak-proof
Diameter (English) 3.5 in.
For Use With (Application) Solution Storage
Includes Screw caps
Height (Metric) With Closure 209.55 mm
Material Polypropylene
Length (Metric) 85 mm
Quantity 1 Ea.
Sterility Non-sterile
Width (Metric) 85 mm
Closure Included Yes
Graduated No
Mouth Narrow
Height (Metric) 220 mm
Height (English) 8.25 in.
Closure Size 20.5 mm
Diameter (Metric) 85 mm
Closure Type Screw Cap
Autoclavable Autoclavable
Capacity (English) 33.8 oz.
Product Type Reagent Bottle
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.