Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Polypropylene Reagent Reservoir
SDPSCI_ED

Catalog No. S01218 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
75 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S01218 75 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S01218 Supplier United Scientific™ Supplier No. 79121
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Reservoir that is durable and affordale for everyday lab usage.

  • For use with micropipettes
  • Total capacity of 75 mL
  • Includes graduation marks at 10, 30, and 60 L levels
  • Autoclavable
  • Sold as a pack of 12

Specifications

Color White
Capacity (Metric) 75 mL
Material Polypropylene
Sterility Non-sterile
Type Reagent Reservoir
For Use With (Application) Everyday Lab Use
Quantity 12/Pk.
Autoclavable Autoclavable
Channels 1
Disposable Reusable
Bottom Shape Flat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.