Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Polypropylene Reversible Reagent Reservoir
SDPSCI_ED

Catalog No. S01219 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Quantity:
6/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S01219 6/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S01219 Supplier United Scientific™ Supplier No. 79131
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Reservoir that is durable and affordale for everyday lab usage.

This versatile polypropylene reservoir is useful for single and multichannel micropipettes.

  • Positioned one way, it provides a 50 mL reservoir for use with a single channel micropipette
  • Flipped over, it provides a numbered reservoir with eight separate 5 mL reagent channels, and a numbered reservoir with twelve separate 5 mL reagent channels
  • Autoclavable
  • Sold as a pack of 6

Specifications

Color White
Capacity (Metric) 50 mL
Material Polypropylene
Sterility Non-sterile
Type Reversible Reservoir
For Use With (Application) Everyday Lab Use
Quantity 6/Pk.
Autoclavable Autoclavable
Channels 8 and 12
Disposable Reusable
Bottom Shape Flat
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.