Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Polypropylene Universal Pipette Tips With Graduation

Catalog No. p-7075522 Shop All Greiner Bio-One Products
Click to view available options
Color:
Natural

Provides optimum performance and fit for all pipettes.

  • Manufactured from high-grade polypropylene
  • Polypropylene has high breaking strength, dimensionally and stable heat-resistant
  • Free of heavy metals
  • Manufactured under production conditions certified conforming to ISO 9001 standards
TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 to 200 μL
Sterility Non-sterile
Color Natural
Autoclavable Autoclavable
Storage Requirements −190° to 140°C
For Use With (Equipment) Universal Fit
Graduated Yes
Material Polypropylene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.