Learn More
Eisco™ Polypropylene Volumetric Flask with Screw Cap
SCI_ED
Catalog No. S24514 Shop All Eisco Products
Change view
Click to view available options
Capacity (Metric):
100 mL
250 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S24514 100 mL
S27345 250 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S24514 Supplier Eisco™ Supplier No. CH0452C
Add to Cart
Add to Cart
Flasks are constructed of high quality, durable polypropylene and fitted with a screw cap.
  • Constructed of strong, high quality polypropylene
  • Single, red graduation mark
  • Fitted with a screw cap
  • Autoclavable for easy sterilization
Specifications
Color Clear
Capacity (Metric) 100 mL
Material Polypropylene
Product Type Volumetric Flask
Autoclavable Autoclavable
Disposable Reusable
Graduated Yes
Mouth Style Narrow
Sterility Non-sterile
Bottom Shape Flat
Closure Type Screw Cap
Surface Treatment Non-treated