Learn More
Polysorbate 80, NF, Spectrum™ Chemical
Shop All Spectrum Chemical Manufacturing Corporation Products

Description
Spectrum™ Chemical Polysorbate 80, NF is used to solubilize essential oils. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
| Acid Value | 2 |
| CAS Max % | 1 |
| pH | Neutral |
| Quantity | 20 L |
| Hydroxyl Value | 65-80 |
| Ignition Residue | 0.0025 |
| Packaging | Amber Glass Bottle |
| Saponification Value | 45-55 |
| Viscosity | 300-500 cSt |
| Water | 0.03 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.