Learn More
United Scientific™ Polystyrene Microscope Slide Tray
SDPSCI_ED
Catalog No. S139544 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Slide Capacity:
20
Catalog No. Slide Capacity
S139544 20
1 options
This item is not returnable. View return policy
Catalog No. S139544 Supplier United Scientific™ Supplier No. 62301
Add to Cart
Add to Cart
This item is not returnable. View return policy
Microscope slide storage tray designed to hold for 20 slides.
  • A unique stepped cutout design simplifies removal of slides and is designed to avoid direct contact of prepared slides with the tray surface
  • Polystyrene trays nest within each other. White color provides good contrast for viewing dyes and stains
Specifications
Type Slide Storage Tray
Slide Capacity 20
Device Holds 3 x 1 in. Microscope Slides
Dimensions (L x W x H) 7.5 x 11.25 x 0.75 in.
Material Polystyrene
Sterility Non-sterile