Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Polystyrene Petri Dishes
SCI_ED

Catalog No. S15094 Shop All Eisco Products
Change view
Click to view available options
Includes:
1 Compartment
2 Compartments
3 Compartments
Catalog No. Includes
S15094 2 Compartments
S15093 1 Compartment
S15095 3 Compartments
3 options

Catalog No. S15094

Supplier: Eisco™ CH2020012

Only null left
Add to Cart
Add to Cart

Disposable, sterile polystyrene petri dishes ideal for microbiology applications.

  • Made of high quality polystyrene
  • Built-in triple vents aid gaseous exchange
  • Multifunctional and ready-to-use
  • Clear and shatter resistant
  • Sterilized in package

Specifications

Autoclavable Not Autoclavable
Bottom Shape Flat
Color Clear
Disposable Limited-use
Gridded Not Gridded
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment Non-treated
Type Petri Dish
Well Count 2
For Use With (Application) Cell Culture
Includes 2 Compartments
Vent Triple
Height (Metric) 15 mm
Diameter (Metric) 100 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.