Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Polystyrene Petri Dishes
SCI_ED

Catalog No. S15095 Shop All Eisco Products
Change view
Click to view available options
Includes:
1 Compartment
2 Compartments
3 Compartments
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes
S15095 3 Compartments
S15093 1 Compartment
S15094 2 Compartments
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S15095 Supplier Eisco™ Supplier No. CH2020013
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Disposable, sterile polystyrene petri dishes ideal for microbiology applications.

  • Made of high quality polystyrene
  • Built-in triple vents aid gaseous exchange
  • Multifunctional and ready-to-use
  • Clear and shatter resistant
  • Sterilized in package

Specifications

Autoclavable Not Autoclavable
Bottom Shape Flat
Color Clear
Disposable Limited-use
Gridded Not Gridded
Material Polystyrene
Shape Round
Sterility Sterile
Surface Treatment Non-treated
Type Petri Dish
Well Count 3
For Use With (Application) Cell Culture
Includes 3 Compartments
Vent Triple
Height (Metric) 1.65 cm
Height (English) 0.65 in.
Diameter (Metric) 100 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.