Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heathrow Scientific™ Polystyrene Sterile Single Wrapped Reagent Reservoir

Catalog No. 03449262
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
25 mL
50 mL
55 mL
100 mL
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
03-449-262 50 mL
03-449-255 5 mL
03-449-257 10 mL
03-449-259 25 mL
03-449-278 55 mL
03-449-266 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 03-449-262 Supplier Heathrow Scientific™ Supplier No. 120644
Add to Cart
Add to Cart
This item is not returnable. View return policy

Reagent Reservoirs offer a fresh and modern design with enhanced features to streamline pipetting procedures.

  • Trough design with a v shaped bottom helps ensure that all the reagent is accessible and can be easily aspirated
  • Thick wall construction and wide base adds stability and durability making the reservoir easy to handle without product deformation which results in less spillage
  • Reservoirs can be conveniently stacked with a unique nesting design that enables easy separation
  • Designed to stay flat on the benchtop and not warp
  • Sterile, sealed in bags and electron beam sterilized
  • DNase, RNase and Pyrogen free
  • Sealed in easy-to-open bags
  • Quality certificate available on request
  • Disposable

Specifications

Capacity (English) 1.7 oz.
Color White
Capacity (Metric) 50 mL
Dimensions (L x W x H) 4 x 1.1 x 1.2 in.
Material Polystyrene
Sterility Sterile
Type Reagent Reservoir
Packaging Single Wrapped
Quantity 1/Bag, 100 Bags/Pk.
Autoclavable Not Autoclavable
Channels 1
Disposable Single-use
Bottom Shape Convoluted
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.