Learn More
Fiber Cell System High flux PS Cartridges, Small, 450cm2
Supplier:  Fiber Cell System C2025D
Encompass_Preferred
20 kD MWCO polysulfone fiber with 450 cm2 of surface area. Only one wrap of oxygenation tubing to reduce system dead volume.
Catalog No. NC1849749
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.