Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Polyvinylpyrrolidone K30, MP Biomedicals™

Catalog No. MP021027861 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP021027861 100 g
MP021027865 500 g
MP210278601 1 kg
MP210278605 5 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MP021027861 Supplier MP Biomedicals, Inc Supplier No. 02102786.1
Add to Cart
Add to Cart

Used as a thickener and as a dispersing agent. Polyvinylpyrrolidone K30, MP Biomedicals is a component of Denhardt's Solution. It is also used as a detoxicant, a complexing agent, a lubricant and a binder.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9003-39-8
Molecular Formula (C6H9NO)n
Molecular Weight (g/mol) 111.14
MDL Number MFCD01076626
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES *-CC(-*)N1CCCC1=O

Specifications

Density 1.23–1.29g/mL (Lit.)
pH 2.8 to 4.2 (5% aq. soln.)
Quantity 100 g
Solubility Information Soluble in water (>100mg/mL, 2.5% aq solution), methanol, ethanol, alcohol, chloroform and glycerol, acetic acid; insoluble in dimethyl ether, ethyl acetate, acetone, toluene, xylene, mineral oil, carbon tetrachloride.
Viscosity 27–35cps (1% aq soln)
Formula Weight 35,000–51,000
Physical Form Powder
Recommended Storage Store at Room Temperature (15–30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.