Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals Pomalidomide

Catalog No. AC465700010
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC465700010 1 g
AC465702500 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC465700010 Supplier Thermo Scientific Chemicals Supplier No. 465700010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 19171-19-8 | C13H11N3O4 | 273.25 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 19171-19-8
Molecular Formula C13H11N3O4
Molecular Weight (g/mol) 273.25
MDL Number MFCD12756407
InChI Key UVSMNLNDYGZFPF-UHFFFAOYNA-N
IUPAC Name 4-amino-2-(2,6-dioxopiperidin-3-yl)-2,3-dihydro-1H-isoindole-1,3-dione
SMILES NC1=CC=CC2=C1C(=O)N(C1CCC(=O)NC1=O)C2=O

Specifications

Melting Point 315.0°C to 320.0°C
Quantity 1 g
Formula Weight 273.24
Chemical Name or Material Pomalidomide
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement Suspected of damaging fertility or the unborn child.

Precautionary Statement GHS P Statement Obtain special instructions before use.

Wear protective gloves/protective clothing/eye protection/face protection.

IF exposed or concerned: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.