Learn More
Medchemexpress LLC HY-130737 25mg Medchemexpress, Pomalidomide 4'-alkylC5-acid CAS:2225940-49-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY13073725MG
Encompass_Preferred
Medchemexpress, HY-130737 25mg Pomalidomide 4'-alkylC5-acid CAS:2225940-49-6 Pomalidomide 4'-alkylC5-acid is a synthesized E3 ligase ligand-linker conjugate that incorporates the Pomalidomide based cereblon ligand and a PEG linker used in PROTAC technology. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1249
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.