Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POMC Monoclonal Antibody (B31.15)
GREENER_CHOICE

Catalog No. 01675931 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-675-931 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-675-931 Supplier Invitrogen™ Supplier No. MA119161
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody reacts with human beta Endorphin, an endogenous opiate derived from ACTH gene. ACTH (Corticotropin; human 39 aa) is synthesized by the anterior pituitary gland and stimulates the adrenal cortex; 6 hormones are derived from one ACTH gene: ACTH, lipotropin, alpha-MSH, beta-MSH, endorphin, and one other. A suggested positive control for this product is human pituitary gland tissue. Immunohistochemistry (Paraffin): Standard ABC technique (DAB+), heat retrieval, incubation: overnight at 4°C.

This gene encodes a polypeptide hormone precursor that undergoes extensive, tissue-specific, post-translational processing via cleavage by subtilisin-like enzymes known as prohormone convertases. There are eight potential cleavage sites within the polypeptide precursor and, depending on tissue type and the available convertases, processing may yield as many as ten biologically active peptides involved in diverse cellular functions. The encoded protein is synthesized mainly in corticotroph cells of the anterior pituitary where four cleavage sites are used; adrenocorticotrophin, essential for normal steroidogenesis and the maintenance of normal adrenal weight, and lipotropin beta are the major end products. In other tissues, including the hypothalamus, placenta, and epithelium, all cleavage sites may be used, giving rise to peptides with roles in pain and energy homeostasis, melanocyte stimulation, and immune modulation. These include several distinct melanotropins, lipotropins, and endorphins that are contained within the adrenocorticotrophin and beta-lipotropin peptides. Mutations in this gene have been associated with early onset obesity, adrenal insufficiency, and red hair pigmentation. Alternatively spliced transcript variants encoding the same protein have been described.
TRUSTED_SUSTAINABILITY

Specifications

Antigen POMC
Applications Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone B31.15
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 15mM sodium azide; pH 7.4
Gene POMC
Gene Accession No. P01189
Gene Alias ACTH; adrenal corticotropic hormone; adrenocorticotropic hormone; adrenocorticotropin; alpha-melanocyte stimulating hormone; alpha-melanocyte-stimulating hormone; alphaMSH; alpha-MSH; BE; beta-endorphin; Beta-LPH; beta-melanocyte-stimulating hormone; beta-MSH; CLIP; Corticotropin; Corticotropin-like intermediary peptide; corticotropin-lipotropin; Gamma-LPH; gamma-MSH; lipotr; Lipotropin beta; lipotropin gamma; LPH; Melanocyte-stimulating hormone alpha; Melanocyte-stimulating hormone beta; Melanotropin alpha; melanotropin beta; Melanotropin gamma; met-enkephalin; MSH; Npp; opiomelanocortin prepropeptide; OTTHUMP00000119991; OTTHUMP00000200964; POC; Pomc; Pomc1; Pomc-1; Pomc2; Potential peptide; Precursor of MSH; pro-ACTH-endorphin; proopiomelanocortin; pro-opiomelanocortin; proopiomelanocortin (adrenocorticotropin/ beta-lipotropin/ alpha-melanocyte stimulating hormone/ beta-melanocyte stimulating hormone/ beta-endorphin); proopiomelanocortin preproprotein; proopiomelanocortin, beta (endorphin, beta); pro-opiomelanocortin-alpha; proopoimelanocortin, beta (endorphin, beta)
Gene Symbols POMC
Host Species Mouse
Immunogen Full length, native human beta Endorphin.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5443
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.