Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ POMT2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114381
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114381 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114381 Supplier Invitrogen™ Supplier No. PA5114381
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Reconstitute with 0.2 mL of distilled water to yield a concentration of 500 μg/mL. Positive Control - WB: human PC-3 whole cell, human Caco-2 whole cell, human HepG2 whole cell, rat lung tissue, rat testis tissue, rat spleen tissue, mouse lung tissue, mouse testis tissue. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.

The protein encoded by this gene is an O-mannosyltransferase that requires interaction with the product of the POMT1 gene for enzymatic function. The encoded protein is found in the membrane of the endoplasmic reticulum. Defects in this gene are a cause of Walker-Warburg syndrome (WWS).
TRUSTED_SUSTAINABILITY

Specifications

Antigen POMT2
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 500 μg/mL
Conjugate Unconjugated
Formulation PBS with 4mg trehalose and 0.05mg sodium azide
Gene POMT2
Gene Accession No. Q8BGQ4, Q9UKY4
Gene Alias A830009D15Rik; AW046274; DKFZp686G10254; Dolichyl-phosphate-mannose--protein mannosyltransferase; dolichyl-phosphate-mannose--protein mannosyltransferase 2; FLJ22309; LGMD2N; MDDGA2; MDDGB2; MDDGC2; Pomt2; protein O-mannosyltransferase 2; Protein O-mannosyl-transferase 2; protein-O-mannosyltransferase 2
Gene Symbols POMT2
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence of human POMT2 (QAARAAGRDVAAEAVARSPKR).
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 217734, 29954, 688673
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.